Ethyl-4-(methylamino)piperidine-1-carboxylate 95%
Supplier: Apollo Scientific
OR911475-25G
OR911475-5G
APOSOR911475-25GEA
539
EUR
APOSOR911475-25G
APOSOR911475-5G
Ethyl-4-(methylamino)piperidine-1-carboxylate 95%
Ethyl-4-(methylamino)piperidine-1-carboxylate
Formula:
C₉H₁₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD07781124 CAS Number: 73733-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...