4-Chloroisatin 97%

Supplier: Apollo Scientific

Synonyms: 4-Chloroindoline-2,3-dione

OR911509-100G OR911509-25G
APOSOR911509-100GEA 200 EUR
APOSOR911509-100G APOSOR911509-25G
4-Chloroisatin 97%
4-Chloroisatin
Formula: C₈H₄ClNO₂
MW: 181,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129178
CAS Number: 6344-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR