7-Ethyl-3-indoleethanol 95%
Supplier: Apollo Scientific
Synonyms:
2-(7-Ethyl-1H-indol-3-yl)ethanol, 7-Ethyltryptophol, 7-Ethyl-3-(2-hydroxyethyl)indole, Etodolac impurity H
OR911544-1G
OR911544-25G
OR911544-5G
APOSOR911544-1GEA
102
EUR
APOSOR911544-1G
APOSOR911544-25G
APOSOR911544-5G
7-Ethyl-3-indoleethanol 95%
7-Ethyl-3-indoleethanol
Formula:
C₁₂H₁₅NO MW: 189,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01718805 CAS Number: 41340-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...