(±)-Tetrahydrofurfuryl chloride 95%

Supplier: Apollo Scientific

OR911546-100G OR911546-25G
APOSOR911546-100GEA 0 EUR
APOSOR911546-100G APOSOR911546-25G
(±)-Tetrahydrofurfuryl chloride 95%
(±)-Tetrahydrofurfuryl chloride
Formula: C₅H₉ClO
MW: 120,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005370
CAS Number: 3003-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR