5,5-Dimethyloxazolidine-2,4-dione (dimethadione) 95%

Supplier: Apollo Scientific

Synonyms: Dimethadione, 5,5-Dimethyloxazolidine-2,4-dione

OR911547-25G OR911547-5G
APOSOR911547-25GEA 61.1 EUR
APOSOR911547-25G APOSOR911547-5G
5,5-Dimethyloxazolidine-2,4-dione (dimethadione) 95%
5,5-Dimethyloxazolidine-2,4-dione (Dimethadione)
Formula: C₅H₇NO₃
MW: 129,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005379
CAS Number: 695-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR