Hinokitiol 95%

Supplier: Apollo Scientific

Synonyms: beta-Thujaplicine, 2-Hydroxy-4-isopropylcyclohepta-2,4,6-trien-1-one

OR911593-25G
APOSOR911593-25GEA 144 EUR
APOSOR911593-25G
Hinokitiol 95%
Hinokitiol
Formula: C₁₀H₁₂O₂
MW: 164,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059582
CAS Number: 499-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR