Methylmaleic hydrazide 98%

Supplier: Apollo Scientific

Synonyms: 3,6-Dihydroxy-4-methylpyridazine, 4-Methyl-3,6-pyridazinediol, 1,2-Dihydro-4-methyl-3,6-pyridazinedione, Methylmaleic Acid Hydrazide

OR911686-100G OR911686-500G
APOSOR911686-100GEA 122 EUR
APOSOR911686-100G APOSOR911686-500G
Methylmaleic hydrazide 98%
Methylmaleic hydrazide
Formula: C₅H₆N₂O₂
MW: 126,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067746
CAS Number: 5754-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR