Boc-D-ABU-OH 97%

Supplier: Apollo Scientific

Synonyms: (R)-2-(Boc-amino)butyric acid

OR911736-25G
APOSOR911736-25GEA 68.3 EUR
APOSOR911736-25G
Boc-D-ABU-OH 97%
Boc-D-ABU-OH
Formula: C₉H₁₇NO₄
MW: 203,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270335
CAS Number: 45121-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR