Boc-N-Me-Phe-OH 97%

Supplier: Apollo Scientific

Synonyms: N-Boc-N-methyl-L-phenylalanine

OR911757-5G
APOSOR911757-5GEA 89.9 EUR
APOSOR911757-5G
Boc-N-Me-Phe-OH 97%
Boc-N-Me-Phe-OH
Formula: C₁₅H₂₁NO₄
MW: 279,336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038754
CAS Number: 37553-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR