2,3,6-Trichloropyridine 98%
Supplier: Apollo Scientific
OR911758-100G
OR911758-25G
APOSOR911758-100GEA
319
EUR
APOSOR911758-100G
APOSOR911758-25G
2,3,6-Trichloropyridine 98%
2,3,6-Trichloropyridine
Formula:
C₅H₂Cl₃N MW: 182,43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00463516 CAS Number: 6515-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...