2-tert-Butylaniline 98%

Supplier: Apollo Scientific

Synonyms: 2-(tert-Butyl)aniline

OR911839-100G
APOSOR911839-100GEA 195 EUR
APOSOR911839-100G
2-tert-Butylaniline 98%
2-tert-Butylaniline
Formula: C₁₀H₁₅N
MW: 149,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130023
CAS Number: 6310-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR