m-Phenetidine 95%
Supplier: Apollo Scientific
Synonyms:
1-Amino-3-ethoxybenzene, 3-Ethoxyaniline
OR911840-25G
OR911840-5G
APOSOR911840-25GEA
435
EUR
APOSOR911840-25G
APOSOR911840-5G
m-Phenetidine 95%
m-Phenetidine
Formula:
C₈H₁₁NO MW: 137,18 g/mol Storage Temperature: Ambient |
CAS Number:
621-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...