2,6-Diethyl-4-methylaniline 98%

Supplier: Apollo Scientific

OR911846-10G OR911846-1G OR911846-5G
APOSOR911846-10GEA 93.1 EUR
APOSOR911846-10G APOSOR911846-1G APOSOR911846-5G
2,6-Diethyl-4-methylaniline 98%
2,6-Diethyl-4-methylaniline
Formula: C₁₁H₁₇N
MW: 163,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035950
CAS Number: 24544-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR