Methyl-4-hydroxy-3-methylbenzoate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 4-hydroxy-3-methylbenzoate

OR911863-10G OR911863-1G OR911863-25G
APOSOR911863-10GEA 364 EUR
APOSOR911863-10G APOSOR911863-1G APOSOR911863-25G
Methyl-4-hydroxy-3-methylbenzoate 98%
Methyl-4-hydroxy-3-methylbenzoate
Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203848
CAS Number: 42113-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR