(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%
Supplier: Apollo Scientific
Synonyms:
(S)-Methyl 2-hydroxy-3-phenylpropanoate, L-β-Phenyllactic acid-OMe
OR912136-100G
APOSOR912136-100GEA
172
EUR
APOSOR912136-100G
(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%
(S)-Methyl-2-hydroxy-3-phenylpropanoate
Formula:
C₁₀H₁₂O₃ MW: 180,2005 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038224 CAS Number: 13673-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...