4(5)-Cyanomethylimidazole 95%

Supplier: Apollo Scientific

OR912312-1G OR912312-5G
APOSOR912312-1GEA 41.6 EUR
APOSOR912312-1G APOSOR912312-5G
4(5)-Cyanomethylimidazole 95%
4(5)-Cyanomethylimidazole
Formula: C₅H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD00070306
CAS Number: 18502-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR