3-Hydroxymethyl-2-methylbiphenyl 98%

Supplier: Apollo Scientific

Synonyms: (2-Methyl-[1,1'-biphenyl]-3-yl)methanol, 2-Methylbiphenyl-3-methanol

OR912371-100G OR912371-25G
APOSOR912371-100GEA 524 EUR
APOSOR912371-100G APOSOR912371-25G
3-Hydroxymethyl-2-methylbiphenyl 98%
3-Hydroxymethyl-2-methylbiphenyl
Formula: C₁₄H₁₄O
MW: 198,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134200
CAS Number: 76350-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR