3-Quinolinol 97%
Supplier: Apollo Scientific
Synonyms:
3-Hydroxyquinoline
OR912414-25G
APOSOR912414-25GEA
114
EUR
APOSOR912414-25G
3-Quinolinol 97%
3-Quinolinol
Formula:
C₉H₇NO MW: 145,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00169018 CAS Number: 580-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...