Ethyl (R)-4-chloro-3-hydroxybutyrate 97%
Supplier: Apollo Scientific
Synonyms:
(R)-Ethyl 4-chloro-3-hydroxybutanoate, (R)-4-Chloro-3-hydroxybutyric Acid Ethyl Ester
OR912444-100G
APOSOR912444-100GEA
82.8
EUR
APOSOR912444-100G
Ethyl (R)-4-chloro-3-hydroxybutyrate 97%
Ethyl (R)-4-chloro-3-hydroxybutyrate
Formula:
C₆H₁₁ClO₃ MW: 166,60 g/mol Boiling Pt: 93…95 °C/5,25mmHg Density: 1,19 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211242 CAS Number: 90866-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...