4-Chloroquinazoline 98%
Supplier: Apollo Scientific
OR912468-25G
APOSOR912468-25GEA
243
EUR
APOSOR912468-25G
4-Chloroquinazoline 98%
4-Chloroquinazoline
Formula:
C₈H₅ClN₂ MW: 164,5900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00228682 CAS Number: 5190-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...