(S)-(+)-Glycidyl butyrate 95%
Supplier: Apollo Scientific
OR912507-10G
OR912507-1G
OR912507-5G
APOSOR912507-10GEA
502
EUR
APOSOR912507-10G
APOSOR912507-1G
APOSOR912507-5G
(S)-(+)-Glycidyl butyrate 95%
(S)-(+)-Glycidyl butyrate
Formula:
C₇H₁₂O₃ MW: 144,1684 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00273369 CAS Number: 65031-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...