3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(BOC-Aminomethyl)benzoic acid, 3-[[(tert-butoxycarbonyl)amino]methyl]benzoic acid, N-Boc-3-Aminomethyl-benzoicacid

OR912508-5G
APOSOR912508-5GEA 66.7 EUR
APOSOR912508-5G
3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid 96%
3-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid
Formula: C₁₃H₁₇NO₄
MW: 251,282 g/mol
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00273426
CAS Number: 117445-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR