4-Acetoxy-2,5-dimethyl-3(2H)-furanone 95%

Supplier: Apollo Scientific

Synonyms: Acetic Acid 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Ester, 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Acetate, Furaneol Acetate, 2,5-Dimethyl-4-oxo-4,5-dihydrofuran-3-yl acetate

OR912574-100G
APOSOR912574-100GEA 88.6 EUR
APOSOR912574-100G
4-Acetoxy-2,5-dimethyl-3(2H)-furanone 95%
4-Acetoxy-2,5-dimethyl-3(2H)-furanone
Formula: C₈H₁₀O₄
MW: 170,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799462
CAS Number: 4166-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR