4-Ethoxypyridin-2-amine 98%

Supplier: Apollo Scientific

Synonyms: 2-Amino-4-ethoxypyridine

OR912692-10G OR912692-1G OR912692-5G
APOSOR912692-10GEA 684 EUR
APOSOR912692-10G APOSOR912692-1G APOSOR912692-5G
4-Ethoxypyridin-2-amine 98%
4-Ethoxypyridin-2-amine
Formula: C₇H₁₀N₂O
MW: 138,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646350
CAS Number: 52311-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR