5-Chloro-1H-indazole-3-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

OR912768-1G OR912768-250MG
APOSOR912768-1GEA 109 EUR
APOSOR912768-1G APOSOR912768-250MG
5-Chloro-1H-indazole-3-carboxylic acid 95%
5-Chloro-1H-indazole-3-carboxylic acid
Formula: C₈H₅ClN₂O₂
MW: 196,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02326712
CAS Number: 1077-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR