Methyl-3-chloropyrazine-2-carboxylate 97%
Supplier: Apollo Scientific
OR912822-1G
OR912822-5G
APOSOR912822-1GEA
24.4
EUR
APOSOR912822-1G
APOSOR912822-5G
Methyl-3-chloropyrazine-2-carboxylate 97%
Methyl-3-chloropyrazine-2-carboxylate
Formula:
C₆H₅ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03093326 CAS Number: 27825-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...