4-Hydroxy-2,5-dimethylbenzaldehyde 95%
Supplier: Apollo Scientific
OR912853-1G
OR912853-5G
APOSOR912853-1GEA
31.6
EUR
APOSOR912853-1G
APOSOR912853-5G
4-Hydroxy-2,5-dimethylbenzaldehyde 95%
4-Hydroxy-2,5-dimethylbenzaldehyde
Formula:
C₉H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03411744 CAS Number: 85231-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...