6-Methylimidazo[1,2-a]pyridine-2-carboxylic acid 98%
Supplier: Apollo Scientific
OR912864-10G
OR912864-1G
OR912864-5G
APOSOR912864-10GEA
267
EUR
APOSOR912864-10G
APOSOR912864-1G
APOSOR912864-5G
6-Methylimidazo[1,2-a]pyridine-2-carboxylic acid 98%
6-Methylimidazo[1,2-a]pyridine-2-carboxylic acid
Formula:
C₉H₈N₂O₂ MW: 176,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425068 CAS Number: 80353-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...