2-Methoxy-5-methylbenzonitrile 95%
Supplier: Apollo Scientific
OR912865-1G
OR912865-5G
APOSOR912865-1GEA
58.2
EUR
APOSOR912865-1G
APOSOR912865-5G
2-Methoxy-5-methylbenzonitrile 95%
2-Methoxy-5-methylbenzonitrile
Formula:
C₉H₉NO MW: 147,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425693 CAS Number: 53078-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...