[2-(Aminomethyl)phenyl]acetic acid 97%
Supplier: Apollo Scientific
OR912943-1G
OR912943-5G
APOSOR912943-1GEA
24.4
EUR
APOSOR912943-1G
APOSOR912943-5G
[2-(Aminomethyl)phenyl]acetic acid 97%
[2-(Aminomethyl)phenyl]acetic acid
Formula:
C₉H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04038440 CAS Number: 40851-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...