Ethyl-2-aminopyrimidine-5-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 2-aminopyrimidine-5-carboxylate, 2-Amino-pyrimidine-5-carboxylic acid ethyl ester

OR912994-5G
APOSOR912994-5GEA 134 EUR
APOSOR912994-5G
Ethyl-2-aminopyrimidine-5-carboxylate 95%
Ethyl-2-aminopyrimidine-5-carboxylate
Formula: C₇H₉N₃O₂
MW: 167,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114100
CAS Number: 57401-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR