N-Methyl-1-(3-nitrophenyl)methanamine 98%
Supplier: Apollo Scientific
Synonyms:
N-Methyl-N-(3-nitrobenzyl)amine
OR913010-10G
OR913010-25G
APOSOR913010-10GEA
183
EUR
APOSOR913010-10G
APOSOR913010-25G
N-Methyl-1-(3-nitrophenyl)methanamine 98%
N-Methyl-1-(3-nitrophenyl)methanamine
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115408 CAS Number: 19499-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...