3-tert-Butylsulphanylpyridine-2-carbonitrile 95%

Supplier: Apollo Scientific

Synonyms: 3-tert-Butylsulphanyl-pyridine-2-carbonitrile

OR913011-1G OR913011-250MG
APOSOR913011-1GEA 435 EUR
APOSOR913011-1G APOSOR913011-250MG
3-tert-Butylsulphanylpyridine-2-carbonitrile 95%
3-tert-Butylsulphanylpyridine-2-carbonitrile
Formula: C₁₀H₁₂N₂S
MW: 192,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116127
CAS Number: 178811-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR