(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95%

Supplier: Apollo Scientific

Synonyms: (R)-(-)-1-(2-Pyrrolidinylmethyl)pyrrolidine

OR913029-1G OR913029-5G
APOSOR913029-1GEA 115 EUR
APOSOR913029-1G APOSOR913029-5G
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95%
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine
Formula: C₉H₁₈N₂
MW: 154,2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117879
CAS Number: 60419-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR