(R)-2-(1-Hydroxyethyl)pyridine 95%

Supplier: Apollo Scientific

Synonyms: (R)-1-(Pyridin-2-yl)ethanol

OR913044-1G OR913044-250MG
APOSOR913044-1GEA 111 EUR
APOSOR913044-1G APOSOR913044-250MG
(R)-2-(1-Hydroxyethyl)pyridine 95%
(R)-2-(1-Hydroxyethyl)pyridine
Formula: C₇H₉NO
MW: 123,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972322
CAS Number: 27911-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR