6-(p-Tolyl)pyridin-3-amine 98%

Supplier: Apollo Scientific

OR913128-1G OR913128-5G
APOSOR913128-1GEA 190 EUR
APOSOR913128-1G APOSOR913128-5G
6-(p-Tolyl)pyridin-3-amine 98%
6-(p-Tolyl)pyridin-3-amine
Formula: C12H12N2
MW: 184,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864839
CAS Number: 170850-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR