5-Methyl-1H-indole-3-carboxylic acid 98%

Supplier: Apollo Scientific

OR913133-10G OR913133-1G OR913133-5G
APOSOR913133-10GEA 285 EUR
APOSOR913133-10G APOSOR913133-1G APOSOR913133-5G
5-Methyl-1H-indole-3-carboxylic acid 98%
5-Methyl-1H-indole-3-carboxylic acid
Formula: C10H9NO2
MW: 175,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203285
CAS Number: 10242-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR