95%
Supplier: Apollo Scientific
OR913389-1G
OR913389-5G
APOSOR913389-1GEA
89.9
EUR
APOSOR913389-1G
APOSOR913389-5G
95%
3-Ethylbenzaldehyde
Formula:
C₉H₁₀O MW: 134.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09263482 CAS Number: 34246-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...