2,6-Dimethyl-4-nitroanisole 96%

Supplier: Apollo Scientific

Synonyms: 2-Methoxy-1,3-dimethyl-5-nitrobenzene

OR913588-1G OR913588-5G
APOSOR913588-1GEA 87.3 EUR
APOSOR913588-1G APOSOR913588-5G
2,6-Dimethyl-4-nitroanisole 96%
2,6-Dimethyl-4-nitroanisole
Formula: C₉H₁₁NO₃
MW: 181,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024537
CAS Number: 14804-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR