(S)-1-(Naphthalen-1-yl)ethanol 96%
Supplier: Apollo Scientific
Synonyms:
(S)-(-)-1-(1-Hydroxyethyl)naphthalene, (S)-(-)-alpha-Methyl-1-naphthalenemethanol, (S)-(-)-1-(1-Naphthyl)ethanol
OR913779-10G
OR913779-1G
OR913779-5G
APOSOR913779-10GEA
267
EUR
APOSOR913779-10G
APOSOR913779-1G
APOSOR913779-5G
(S)-1-(Naphthalen-1-yl)ethanol 96%
(S)-1-(Naphthalen-1-yl)ethanol
Formula:
C₁₂H₁₂O MW: 172,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077831 CAS Number: 15914-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...