2-Bromo-5-cyano-3-picoline 95%

Supplier: Apollo Scientific

OR914260-1G OR914260-250MG
APOSOR914260-1GEA 109 EUR
APOSOR914260-1G APOSOR914260-250MG
2-Bromo-5-cyano-3-picoline 95%
2-Bromo-5-cyano-3-picoline
Formula: C₇H₅BrN₂
MW: 197,0320 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095063
CAS Number: 374633-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR