4-Vinyl-1,3-dioxolan-2-one 98%

Supplier: Apollo Scientific

OR914384-100G OR914384-500G
APOSOR914384-100GEA 63.8 EUR
APOSOR914384-100G APOSOR914384-500G
4-Vinyl-1,3-dioxolan-2-one 98%
4-Vinyl-1,3-dioxolan-2-one
Formula: C₅H₆O₃
MW: 114,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143315
CAS Number: 4427-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR