2-Hydroxy-2-(4-methylphenyl)acetic acid 98%
Supplier: Apollo Scientific
OR914401-25G
OR914401-5G
APOSOR914401-25GEA
509
EUR
APOSOR914401-25G
APOSOR914401-5G
2-Hydroxy-2-(4-methylphenyl)acetic acid 98%
2-Hydroxy-2-(4-methylphenyl)acetic acid
Formula:
C₉H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00961860 CAS Number: 31284-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...