3,4-Dihydro-2H-1,5-benzodioxepin 97%

Supplier: Apollo Scientific

OR914681-1G OR914681-5G
APOSOR914681-1GEA 0 EUR
APOSOR914681-1G APOSOR914681-5G
3,4-Dihydro-2H-1,5-benzodioxepin 97%
3,4-Dihydro-2H-1,5-benzodioxepin
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD01333457
CAS Number: 7216-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR