(±)-Methyl mandelate 95%
Supplier: Apollo Scientific
Synonyms:
DL-Methyl glycolate, DL-2-Hydroxy-2-phenylacetic acid methyl ester, (±)-2-Hydroxy-2-phenylacetic acid methyl ester, (±)-Mandelic acid methyl ester, Methyl DL-mandelate, Methyl 2-hydroxy-2-phenylacetate, (±)-Methyl glycolate, DL-Mandelic acid methyl ester, DL-Methyl mandelate
OR914697-100G
OR914697-25G
APOSOR914697-100GEA
81.5
EUR
APOSOR914697-100G
APOSOR914697-25G
(±)-Methyl mandelate 95%
(±)-Methyl mandelate
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004493 CAS Number: 4358-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...