4-Phenylazophenol 95%

Supplier: Apollo Scientific

Synonyms: 4-(Phenyldiazenyl)phenol

OR916223-100G OR916223-250G OR916223-25G
APOSOR916223-100GEA 285 EUR
APOSOR916223-100G APOSOR916223-250G APOSOR916223-25G
4-Phenylazophenol 95%
4-Phenylazophenol
Formula: C₁₂H₁₀N₂O
MW: 198,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002330
CAS Number: 1689-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR