5-Amino-1-phenyl-1H-pyrazole 98%
Supplier: Apollo Scientific
Synonyms:
1-Phenyl-1H-pyrazol-5-amine
OR916328-1G
OR916328-5G
APOSOR916328-1GEA
147
EUR
APOSOR916328-1G
APOSOR916328-5G
5-Amino-1-phenyl-1H-pyrazole 98%
5-Amino-1-phenyl-1H-pyrazole
Formula:
C₉H₉N₃ MW: 159,1900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00020734 CAS Number: 826-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...