Propyl lactate 95%

Supplier: Apollo Scientific

OR916585-25G OR916585-5G
APOSOR916585-25GEA 183 EUR
APOSOR916585-25G APOSOR916585-5G
Propyl lactate 95%
Propyl lactate
Formula: C₆H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00059661
CAS Number: 616-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR