5-Chloro-2-methyl-1,3-benzoxazol-6-amine 98%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-5-chloro-6-benzoxazolamine

OR916745-1G OR916745-5G
APOSOR916745-1GEA 101 EUR
APOSOR916745-1G APOSOR916745-5G
5-Chloro-2-methyl-1,3-benzoxazol-6-amine 98%
5-Chloro-2-methyl-1,3-benzoxazol-6-amine
Formula: C₈H₇ClN₂O
MW: 182,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368636
CAS Number: 323579-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR