H-DL-Phe(4-Me)-OH 97%
Supplier: Apollo Scientific
Synonyms:
4-Methyl-DL-phenylalanine
OR916821-1G
OR916821-25G
OR916821-5G
APOSOR916821-1GEA
118
EUR
APOSOR916821-1G
APOSOR916821-25G
APOSOR916821-5G
H-DL-Phe(4-Me)-OH 97%
H-DL-Phe(4-Me)-OH
Formula:
C₁₀H₁₃NO₂ MW: 179,2 g/mol Storage Temperature: Ambient |
CAS Number:
4599-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...